| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015047.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015047.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015047.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015047.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015047.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015047.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212104.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212104.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212104.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212104.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212104.hg.1 |
TC0100015047.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015047.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015047.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015047.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015047.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015047.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015047.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212105.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212105.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212105.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212105.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212105.hg.1 |
TC0100015047.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100212103.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212103.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212103.hg.1 |
ENST00000438829.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212103.hg.1 |
NR_125951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212103.hg.1 |
TC0100015047.hg.1 |
Clariom D TC |