| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100224596.hg.1 |
TC0100016377.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000416416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000421020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000446102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016377.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100224605.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224605.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224605.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100224605.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224605.hg.1 |
ENST00000456083.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100224605.hg.1 |
NR_125958.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100224605.hg.1 |
TC0100016377.hg.1 |
Clariom D TC |