Assay Details
Gene Symbol: LOC101928673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125960.1 | 1 - 2 | 988 | 68 | ||
GenBank mRNA | AK130460.1 | - | 1 - 2 | 989 | 68 | |
BC136808.1 | - | 1 - 2 | 981 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100506023 | LOC100506023 | Sense | Chr. 1 - 173235060 - 173477155 on NCBI Build GRCh38 | ||
Downstream | peroxiredoxin 6 | PRDX6 |
1-Cys, AOP2, HEL-S-128m, NSGPx, PRX, aiPLA2, p29 |
16.0kb | Anti-sense | Chr. 1 - 173477347 - 173488807 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224851.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224851.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224851.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224851.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224851.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224841.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224841.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224841.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224841.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224841.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224841.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224841.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224852.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224852.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224852.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224852.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224852.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224853.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224853.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224853.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224853.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224853.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224854.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224854.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224854.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224854.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224854.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | ENST00000437190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_037845.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016432.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224856.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224856.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224856.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224856.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224856.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224850.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224850.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224850.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224850.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224850.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224855.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224855.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224855.hg.1 | ENST00000367716.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224855.hg.1 | NR_125960.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224855.hg.1 | TC0100016432.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 108162 | - | - |
