Assay Details
| Gene Symbol: LOC101928977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125970.1 | 1 - 2 | 176 | 86 | ||
| GenBank mRNA | DA284216.1 | - | 1 - 2 | 258 | 86 | |
| DA297063.1 | - | 1 - 2 | 177 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928995 | LOC101928995 | -33.6kb | Anti-sense | Chr. 1 - 115919376 - 115925908 on NCBI Build GRCh38 | |
| Downstream | mab-21 like 3 | MAB21L3 | C1orf161 | 98.5kb | Anti-sense | Chr. 1 - 116072744 - 116135240 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928977
Genomic Map