Assay Details
| Gene Symbol: LOC101929023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125972.1 | 1 - 2 | 51 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATP1A1 antisense RNA 1 | ATP1A1-AS1 |
ATP1A1OS, C1orf203 |
-59.4kb | Sense | Chr. 1 - 116392865 - 116418622 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105378922 | LOC105378922 | 14.2kb | Sense | Chr. 1 - 116493016 - 116505088 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215039.hg.1 | TC0100015394.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000430316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000437308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015394.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000423907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000434879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | ENST00000443219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | NR_125972.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | NR_125973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215043.hg.1 | TC0100015394.hg.1 | Clariom D TC |

LOC101929023
Genomic Map