Assay Details
Gene Symbol: LOC101929626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125987.1 | 2 - 3 | 138 | 122 | ||
GenBank mRNA | AA993332.1 | - | 1 - 2 | 122 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microtubule associated serine/threonine kinase 2 | MAST2 |
MAST205, MTSSK |
-98.4kb | Sense | Chr. 1 - 45786992 - 46036124 on NCBI Build GRCh38 |
Downstream | tetraspanin 1 | TSPAN1 |
NET1, TM4C, TM4SF |
36.0kb | Sense | Chr. 1 - 46175077 - 46196489 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
