Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600183020.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183020.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600183020.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183020.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600183020.hg.1 |
TC1600010746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600183029.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183029.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600183029.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183029.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600183029.hg.1 |
TC1600010746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600183028.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183028.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183028.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600183028.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183028.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600183028.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600183028.hg.1 |
TC1600010746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000502126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
ENST00000566989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010746.hg.1 |
NR_126007.1 |
Refseq |