Assay Details
| Gene Symbol: LOC388406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126009.1 | 1 - 2 | 81 | 115 | ||
| GenBank mRNA | AK131012.1 | - | 1 - 2 | 82 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | amyloid beta precursor protein binding protein 2 | APPBP2 |
APP-BP2, HS.84084, PAT1 |
-38.3kb | Anti-sense | Chr. 17 - 60443149 - 60526240 on NCBI Build GRCh38 |
| Downstream |
protein phosphatase, Mg2+/Mn2+ dependent 1D |
PPM1D |
PP2C-DELTA, WIP1 |
34.7kb | Sense | Chr. 17 - 60600183 - 60666280 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC388406
Genomic Map