Assay Details
| Gene Symbol: LOC100128317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126026.1 | 5 - 6 | 642 | 103 | ||
| GenBank mRNA | AY927634.1 | - | 5 - 6 | 642 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105369146 | LOC105369146 | -16.6kb | Sense | Chr. 7 - 81515747 - 81561819 on NCBI Build GRCh38 | |
| Downstream | hepatocyte growth factor | HGF |
DFNB39, F-TCF, HGFB, HPTA, SF |
104.4kb | Sense | Chr. 7 - 81699006 - 81770438 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183002.hg.1 | TC0700011640.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000413944.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011640.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183003.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183003.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183003.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183003.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183003.hg.1 | ENST00000455420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183003.hg.1 | NR_126026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183003.hg.1 | TC0700011640.hg.1 | Clariom D TC |

LOC100128317
Genomic Map