Assay Details
| Gene Symbol: GLYCTK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046719.1 | 1 | 1025 | 99 | ||
| GenBank mRNA | AI174658.1 | - | 1 | 99 | ||
| BM672046.1 | - | 1 | 99 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycerate kinase | GLYCTK |
HBEBP2, HBEBP4, HBeAgBP4A |
Anti-sense | Chr. 3 - 52287558 - 52295256 on NCBI Build GRCh38 | |
| Downstream | microRNA 135a-1 | MIR135A1 |
MIRN135-1, MIRN135A1, mir-135a-1 |
4.5kb | Sense | Chr. 3 - 52294219 - 52294308 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

GLYCTK-AS1
Genomic Map