| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007166.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007166.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007166.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007166.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007166.hg.1 |
NR_046742.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007166.hg.1 |
NR_046742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148835.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148835.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148835.hg.1 |
NR_046742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148835.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148835.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148835.hg.1 |
NR_046742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148835.hg.1 |
TC0X00007166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00148836.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148836.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148836.hg.1 |
NR_046742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148836.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148836.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148836.hg.1 |
NR_046742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148836.hg.1 |
TC0X00007166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007166.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007166.hg.1 |
ENST00000436124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007166.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007166.hg.1 |
ENST00000614448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007166.hg.1 |
NR_046742.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007166.hg.1 |
NR_046742.2 |
Refseq |