Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161616.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161616.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161616.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161616.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161616.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161616.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161616.hg.1 |
TC1300009144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
ENST00000444536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
ENST00000451570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009144.hg.1 |
NR_047011.1 |
Refseq |