Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009644.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009644.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009644.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009644.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009644.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009644.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164469.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164469.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164469.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164469.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164469.hg.1 |
TC1300009644.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009644.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009644.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009644.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009644.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009644.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009644.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164468.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164468.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164468.hg.1 |
ENST00000436722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164468.hg.1 |
NR_047015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164468.hg.1 |
TC1300009644.hg.1 |
Clariom D TC |