| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009366.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009366.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009366.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009366.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009366.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009366.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009366.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009366.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169254.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169254.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169254.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169254.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169254.hg.1 |
TC1400009366.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400169255.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169255.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169255.hg.1 |
ENST00000557544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169255.hg.1 |
NR_047116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169255.hg.1 |
TC1400009366.hg.1 |
Clariom D TC |