Assay Details
| Gene Symbol: ONECUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073510.1 | 1 - 2 | 287 | 82 | ||
| GenBank mRNA | BX118406.1 | - | 1 - 2 | 287 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 214 member A | FAM214A | KIAA1370 | -47.1kb | Sense | Chr. 15 - 52581321 - 52709817 on NCBI Build GRCh38 |
| Downstream | WD repeat domain 72 | WDR72 | AI2A3 | 732.9kb | Sense | Chr. 15 - 53513741 - 53762878 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
liver development
regulation of cell-matrix adhesion
epithelial cell development
glucose metabolic process
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
endoderm development
cell differentiation
B cell differentiation
positive regulation of cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
endocrine pancreas development
cilium assembly
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
spleen development
system development
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
liver development
regulation of cell-matrix adhesion
epithelial cell development
glucose metabolic process
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
endoderm development
cell differentiation
B cell differentiation
positive regulation of cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
endocrine pancreas development
cilium assembly
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
spleen development
system development
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169724.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169724.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169724.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169724.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169724.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169724.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169724.hg.1 | TC1500009466.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169726.hg.1 | TC1500009466.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169725.hg.1 | TC1500009466.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500169723.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169723.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169723.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169723.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169723.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169723.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169723.hg.1 | TC1500009466.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500169722.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169722.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169722.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169722.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169722.hg.1 | TC1500009466.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500169728.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169728.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169728.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500169728.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500169728.hg.1 | TC1500009466.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000305901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000560699.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000561401.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | ENST00000570208.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NM_004498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NR_073510.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009466.hg.1 | NR_073510.2 | Refseq |

ONECUT1
Genomic Map