Assay Details
| Gene Symbol: LINC00371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102431.2 | 3 - 4 | 308 | 83 | ||
| NR_102432.2 | 2 - 3 | 377 | 83 | |||
| GenBank mRNA | CX751673.1 | - | 308 | 83 | ||
| DR156254.1 | - | 2 - 3 | 377 | 83 | ||
| DV080333.1 | - | 2 - 3 | 268 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | guanylate cyclase 1 soluble subunit beta 2 (pseudogene) | GUCY1B2 | GC-SB2 | -19.2kb | Sense | Chr. 13 - 50994511 - 51066157 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 124 member A | FAM124A | 134.3kb | Anti-sense | Chr. 13 - 51222328 - 51284241 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000507929.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000519248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000520543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000521523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008590.hg.1 | ENST00000523682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160576.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160576.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160576.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160576.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160576.hg.1 | TC1300008973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |

LINC00371
Genomic Map