Assay Details
| Gene Symbol: XXYLT1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102710.1 | 1 - 2 | 233 | 98 | ||
| GenBank mRNA | AW963078.1 | - | 1 - 2 | 208 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3137 | MIR3137 | -13.3kb | Anti-sense | Chr. 3 - 195134506 - 195134580 on NCBI Build GRCh38 | |
| Downstream |
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 |
ACAP2 |
CENTB2, CNT-B2 |
121.9kb | Anti-sense | Chr. 3 - 195274736 - 195443176 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

XXYLT1-AS2
Genomic Map