Assay Details
| Gene Symbol: LOC101593348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103497.1 | 1 | 1920 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4 | B3GNT4 |
B3GN-T4, beta3Gn-T4 |
-18.3kb | Sense | Chr. 12 - 122203681 - 122208135 on NCBI Build GRCh38 |
| Downstream |
VPS33A, CORVET/HOPS core subunit |
VPS33A | 2.8kb | Anti-sense | Chr. 12 - 122231547 - 122266521 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101593348
Genomic Map