Assay Details
| Gene Symbol: NKX2-1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103710.1 | 1 - 2 | 404 | 64 | ||
| GenBank mRNA | BX161496.1 | - | 1 - 2 | 366 | 64 | |
| DA256249.1 | - | 1 - 2 | 404 | 64 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BX161496.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006931.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006931.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006931.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006931.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006931.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006931.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148146.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148146.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148146.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148146.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148146.hg.1 | TC1400006931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400148148.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148148.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148148.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148148.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148148.hg.1 | TC1400006931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000354822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000498187.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000518149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000522719.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | ENST00000546983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_001079668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010734.hg.1 | NM_003317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148147.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148147.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148147.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148147.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148147.hg.1 | TC1400006931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006931.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006931.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006931.hg.1 | ENST00000521292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006931.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006931.hg.1 | NR_103710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006931.hg.1 | NR_103710.1 | Refseq |

NKX2-1-AS1
Genomic Map