Assay Details
| Gene Symbol: KLF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103759.1 | 2 - 3 | 730 | 69 | ||
| GenBank mRNA | HF546211.1 | - | 2 - 3 | 422 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | outer dense fiber of sperm tails 1 | ODF1 |
CT133, HSPB10, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, RT7, SODF |
-87.8kb | Anti-sense | Chr. 8 - 102551620 - 102561017 on NCBI Build GRCh38 |
| Downstream | vegetative cell wall protein gp1-like | LOC101927245 | 1.9kb | Anti-sense | Chr. 8 - 102654313 - 102730620 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

KLF10
Genomic Map