| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144690.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144690.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144690.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144690.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144690.hg.1 |
TC1300006555.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144695.hg.1 |
TC1300006555.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144693.hg.1 |
TC1300006555.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300144696.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144696.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144696.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144696.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144696.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144696.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144696.hg.1 |
TC1300006555.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144694.hg.1 |
TC1300006555.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |