Assay Details
| Gene Symbol: ANTXRLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103827.1 | 4 - 5 | 486 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ArfGAP with GTPase domain, ankyrin repeat and PH domain 7, pseudogene |
AGAP7P |
AGAP-7, AGAP7, CTGLF4, bA109G10.1 |
-87.3kb | Anti-sense | Chr. 10 - 46109495 - 46131678 on NCBI Build GRCh38 |
| Downstream | basic proline-rich protein-like | LOC105378577 | 34.3kb | Anti-sense | Chr. 10 - 46283571 - 46285997 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

ANTXRLP1
Genomic Map