Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188335.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188335.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188335.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188335.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188335.hg.1 |
TC1100011461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100188338.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188338.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188338.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188338.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188338.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188338.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188338.hg.1 |
TC1100011461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100188336.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188336.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188336.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188336.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188336.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188336.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188336.hg.1 |
TC1100011461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100188340.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188340.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188340.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188340.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188340.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188340.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188340.hg.1 |
TC1100011461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000531347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
ENST00000533327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011461.hg.1 |
NR_103835.1 |
Refseq |