Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187657.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187657.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187657.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187657.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187657.hg.1 |
TC0800012441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800187658.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187658.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187658.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187658.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187658.hg.1 |
TC0800012441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800187660.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187660.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187660.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187660.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187660.hg.1 |
TC0800012441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012441.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187655.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187655.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187655.hg.1 |
TC0800012441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800187656.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187656.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187656.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187656.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187656.hg.1 |
TC0800012441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800187659.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187659.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187659.hg.1 |
ENST00000517505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187659.hg.1 |
NR_104062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187659.hg.1 |
TC0800012441.hg.1 |
Clariom D TC |