| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012800.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012800.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012800.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012800.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012800.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188269.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188269.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188269.hg.1 |
TC0600012800.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600188272.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188272.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188272.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188272.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188272.hg.1 |
TC0600012800.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600188271.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188271.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188271.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600188271.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600188271.hg.1 |
TC0600012800.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600012800.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012800.hg.1 |
ENST00000426737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012800.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012800.hg.1 |
NR_104137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012800.hg.1 |
NR_104137.1 |
Refseq |