| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000602425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000602425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
ENST00000602425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000602425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000602425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
ENST00000602425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011135.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186412.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186412.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186412.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186412.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186412.hg.1 |
TC1600011135.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600186413.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186413.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186413.hg.1 |
ENST00000597373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186413.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186413.hg.1 |
TC1600011135.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600186414.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186414.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186414.hg.1 |
TC1600011135.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600186415.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186415.hg.1 |
NR_104139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186415.hg.1 |
TC1600011135.hg.1 |
Clariom D TC |