Assay Details
| Gene Symbol: LINC00458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108063.1 | 3 | 720 | 129 | ||
| GenBank mRNA | BC017942.1 | - | 3 | 715 | 129 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 558 | LINC00558 | -250.7kb | Anti-sense | Chr. 13 - 53815419 - 53876119 on NCBI Build GRCh38 | |
| Downstream | microRNA 1297 | MIR1297 |
MIRN1297, hsa-mir-1297, mir-1297 |
184.5kb | Sense | Chr. 13 - 54311972 - 54312048 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300161176.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300161176.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300161176.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300161176.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300161176.hg.1 | TC1300009042.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000423442.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000427299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000451744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000606706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | ENST00000607494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108063.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009042.hg.1 | NR_108063.1 | Refseq |

LINC00458
Genomic Map