Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012502.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185709.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185709.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185709.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185709.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185709.hg.1 |
TC0400012502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400185712.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185712.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185712.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185712.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185712.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185712.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185712.hg.1 |
TC0400012502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012502.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185707.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185707.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185707.hg.1 |
TC0400012502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400185708.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185708.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185708.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185708.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185708.hg.1 |
TC0400012502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
ENST00000507011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
ENST00000512527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
ENST00000514736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
NR_108092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
NR_108093.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185710.hg.1 |
TC0400012502.hg.1 |
Clariom D TC |