| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159057.hg.1 |
TC0300008303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300159059.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159059.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159059.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159059.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159059.hg.1 |
TC0300008303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300159056.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159056.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159056.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159056.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159056.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159056.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159056.hg.1 |
TC0300008303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300159060.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159060.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159060.hg.1 |
TC0300008303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
ENST00000489670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
ENST00000497996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
NR_109841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
NR_109842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300159058.hg.1 |
TC0300008303.hg.1 |
Clariom D TC |