Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
NR_110625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009374.hg.1 |
NR_110625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
NR_110625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
TC2000009374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165580.hg.1 |
NR_110625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165581.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165581.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165581.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165581.hg.1 |
TC2000009374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000165581.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165581.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165581.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
NR_110625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009374.hg.1 |
NR_110625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
NR_110625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
TC2000009374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
ENST00000417781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
ENST00000428190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
NR_110624.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000165584.hg.1 |
NR_110625.1 |
Refseq |