Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149363.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149363.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149363.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149363.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149363.hg.1 |
TC1500007017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007017.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149360.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149360.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149360.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149360.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149360.hg.1 |
TC1500007017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500149365.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149365.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149365.hg.1 |
TC1500007017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500149359.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149359.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149359.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149359.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149359.hg.1 |
TC1500007017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500149364.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149364.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149364.hg.1 |
TC1500007017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500149361.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149361.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149361.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149361.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149361.hg.1 |
TC1500007017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500149362.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149362.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149362.hg.1 |
ENST00000564168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500149362.hg.1 |
NR_120332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500149362.hg.1 |
TC1500007017.hg.1 |
Clariom D TC |