| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
NR_120533.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009459.hg.1 |
NR_120533.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171181.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171181.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171181.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171181.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171181.hg.1 |
TC1100009459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171180.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171180.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171180.hg.1 |
TC1100009459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
NR_120533.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
NR_120533.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171183.hg.1 |
TC1100009459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171182.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171182.hg.1 |
NR_120533.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171182.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171182.hg.1 |
NR_120533.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171182.hg.1 |
TC1100009459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
ENST00000534620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
ENST00000550747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
NR_120532.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
NR_120533.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009459.hg.1 |
NR_120533.1 |
Refseq |