Assay Details
| Gene Symbol: LYRM4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126016.1 | 3 | 1195 | 129 | ||
| NR_126017.1 | 3 | 1113 | 129 | |||
| NR_126018.1 | 2 | 1100 | 129 | |||
| NR_126019.1 | 2 | 1182 | 129 | |||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AI473891.1 | - | 1 | Chr. 6 - 5043070 - 5043589 [-] on Build GRCh38 | 129 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LYR motif containing 4 | LYRM4 |
C6orf149, CGI-203, COXPD19, ISD11 |
Anti-sense | Chr. 6 - 5031029 - 5260950 on NCBI Build GRCh38 | |
| Downstream | protein phosphatase 1 regulatory subunit 3G | PPP1R3G | 41.9kb | Sense | Chr. 6 - 5085486 - 5087221 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000606767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146704.hg.1 | TC0600006803.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146707.hg.1 | TC0600006803.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000606767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | ENST00000607327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006803.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | ENST00000606761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126017.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126018.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | NR_126019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146703.hg.1 | TC0600006803.hg.1 | Clariom D TC |

LYRM4-AS1
Genomic Map