Assay Details
| Gene Symbol: LOC101927188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126040.1 | 2 | 2019 | 74 | ||
| GenBank mRNA | BC041471.1 | - | 2 | 2019 | 74 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DA216077.1 | - | 1 | Chr. 18 - 6956882 - 6957362 [-] on Build GRCh38 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 668 | LINC00668 | -25.5kb | Anti-sense | Chr. 18 - 6925474 - 6929869 on NCBI Build GRCh38 | |
| Downstream | leucine rich repeat containing 30 | LRRC30 | 273.7kb | Sense | Chr. 18 - 7231139 - 7232044 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927188
Genomic Map