Assay Details
| Gene Symbol: ALMS1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046762.1 | 2 | 417 | 117 | ||
| GenBank mRNA | BC041487.1 | - | 2 | 424 | 117 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM451564.1 | - | 423 | Chr. - - [-] on Build | 117 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ALMS1, centrosome and basal body associated protein |
ALMS1 | ALSS | Sense | Chr. 2 - 73385758 - 73609919 on NCBI Build GRCh38 | |
| Downstream | N-acetyltransferase 8 (putative) | NAT8 |
ATase2, CCNAT, CML1, GLA, Hcml1, TSC501, TSC510 |
181.2kb | Anti-sense | Chr. 2 - 73640723 - 73642410 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ALMS1-IT1
Genomic Map