| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013126.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013126.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013126.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013126.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013126.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013126.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198187.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198187.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198187.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198187.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198187.hg.1 |
TC0300013126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000334567.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000420375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000450693.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000469981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
ENST00000619900.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009497.hg.1 |
NM_001164436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198185.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198185.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198185.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198185.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198185.hg.1 |
TC0300013126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300198186.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198186.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198186.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198186.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198186.hg.1 |
TC0300013126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013126.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013126.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013126.hg.1 |
ENST00000449106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013126.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013126.hg.1 |
NR_046852.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013126.hg.1 |
NR_046852.1 |
Refseq |