Assay Details
| Gene Symbol: CSAG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073432.1 | 4 | 521 | 79 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BF791772.1 | - | 521 | Chr. - - [-] on Build | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAGE family member A12 | MAGEA12 |
CT1.12, MAGE12 |
-2.9kb | Sense | Chr. X - 152733779 - 152737669 on NCBI Build GRCh38 |
| Downstream | MAGE family member A2 | MAGEA2 |
CT1.2, MAGE2, MAGEA2A |
8.9kb | Anti-sense | Chr. X - 152749863 - 152753942 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000357916.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000393900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_001166387.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NM_005367.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008752.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159552.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159552.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159552.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159552.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159552.hg.1 | TC0X00008752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159556.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159556.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159556.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159556.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159556.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159556.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159556.hg.1 | TC0X00008752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159555.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159555.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159555.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159555.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159555.hg.1 | ENST00000583763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159555.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159555.hg.1 | TC0X00008752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159557.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159557.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159557.hg.1 | ENST00000361201.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159557.hg.1 | NR_073432.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159557.hg.1 | TC0X00008752.hg.1 | Clariom D TC |

CSAG4
Genomic Map