Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000582805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000582805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162549.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162549.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162549.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162549.hg.1 |
TC1800009138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162549.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162549.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162549.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162548.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162548.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162548.hg.1 |
TC1800009138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162548.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162548.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000577408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000578833.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000582805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000582805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
ENST00000583106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009138.hg.1 |
NR_104127.1 |
Refseq |