Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186907.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186907.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186907.hg.1 |
TC0600012608.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186910.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186910.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186910.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186910.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186910.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186910.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186910.hg.1 |
TC0600012608.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186909.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186909.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186909.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186909.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186909.hg.1 |
TC0600012608.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012608.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186906.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186906.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186906.hg.1 |
TC0600012608.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186911.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186911.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186911.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186911.hg.1 |
ENST00000564541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186911.hg.1 |
NR_047502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186911.hg.1 |
NR_104136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186911.hg.1 |
TC0600012608.hg.1 |
Clariom D TC |