| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011305.hg.1 |
ENST00000603878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011305.hg.1 |
ENST00000603878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010184.hg.1 |
ENST00000598912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010184.hg.1 |
ENST00000598912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010184.hg.1 |
ENST00000598912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010184.hg.1 |
ENST00000598912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196935.hg.1 |
NR_104148.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196935.hg.1 |
NR_104149.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196935.hg.1 |
NR_104148.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196935.hg.1 |
NR_104149.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196935.hg.1 |
TC0700013370.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700011305.hg.1 |
ENST00000603878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011305.hg.1 |
ENST00000603878.1 |
Ensembl |