Assay Details
| Gene Symbol: MIR222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029636.1 | 1 | 46 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 221 | MIR221 |
MIRN221, miRNA221, mir-221 |
-749.0 | Sense | Chr. X - 45746157 - 45746266 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC401585 | LOC401585 | 101.0kb | Sense | Chr. X - 45848074 - 45851490 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00011361.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000986.hg.1
Set Membership: |
|

MIR222
Genomic Map