Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011279.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011279.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011279.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011279.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011279.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011279.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178192.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178192.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178192.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178192.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178192.hg.1 |
TC0500011279.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500178194.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178194.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178194.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178194.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178194.hg.1 |
TC0500011279.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011279.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011279.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011279.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011279.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011279.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011279.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178193.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178193.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178193.hg.1 |
ENST00000515871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178193.hg.1 |
NR_125754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178193.hg.1 |
TC0500011279.hg.1 |
Clariom D TC |