Assay Details
| Gene Symbol: MIR6127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106742.1 | 1 | 53 | 56 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EPH receptor A8 | EPHA8 |
EEK, EK3, HEK3 |
-29.7kb | Anti-sense | Chr. 1 - 22563508 - 22603594 on NCBI Build GRCh38 |
| Downstream | complement C1q A chain | C1QA | 3.3kb | Anti-sense | Chr. 1 - 22636625 - 22639682 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100013255.hg.1
Set Membership: |
|

MIR6127
Genomic Map