Assay Details
| Gene Symbol: MIR6745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106803.1 | 1 | 72 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP ribosylation factor GTPase activating protein 2 | ARFGAP2 |
IRZ, NBLA10535, ZFP289, ZNF289 |
-2.1kb | Sense | Chr. 11 - 47164298 - 47177503 on NCBI Build GRCh38 |
| Downstream | damage specific DNA binding protein 2 | DDB2 |
DDBB, UV-DDB2, XPE |
35.2kb | Anti-sense | Chr. 11 - 47214942 - 47239218 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1100010727.hg.1
Set Membership: |
|

MIR6745
Genomic Map