Assay Details
| Gene Symbol: BAIAP2L1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BAIAP2L1-MET_B9M15.0 | - | 69 | 118 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAIAP2L1-MET fusion| Gene Symbol: BAIAP2L1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BAIAP2L1-MET_B9M15.0 | - | 69 | 118 | ||
Set Membership: |
|