Assay Details
| Gene Symbol: CEL-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CEL-NTRK1_C7N7_1.0 | - | 61 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEL-NTRK1 fusion| Gene Symbol: CEL-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CEL-NTRK1_C7N7_1.0 | - | 61 | 99 | ||