Assay Details
| Gene Symbol: CLIP4-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLIP4-ALK_C12A23.0 | - | 90 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLIP4-ALK fusion| Gene Symbol: CLIP4-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLIP4-ALK_C12A23.0 | - | 90 | 99 | ||