Assay Details
| Gene Symbol: FGFR3-TACC3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR3-TACC3_F17Intron17T9.0 | - | 30 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR3-TACC3 fusion| Gene Symbol: FGFR3-TACC3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR3-TACC3_F17Intron17T9.0 | - | 30 | 90 | ||