Assay Details
| Gene Symbol: NACC2-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NACC2-NTRK2_N4N13.0 | - | 105 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NACC2-NTRK2 fusion| Gene Symbol: NACC2-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NACC2-NTRK2_N4N13.0 | - | 105 | 67 | ||
Set Membership: |
|