Assay Details
| Gene Symbol: RANBP2-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RANBP2-ALK_R18A20.0 | - | 74 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RANBP2-ALK fusion| Gene Symbol: RANBP2-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RANBP2-ALK_R18A20.0 | - | 74 | 106 | ||
Set Membership: |
|